missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RTD Pt 100 Temperature Probe for Temp 16 Meter

Temperature probes are used to measure values in various educational, laboratory, and industrial applications.

Brand:  Thermo Scientific™ PT56/500

 View more versions of this product

Product Code. 10626793

  • 166.00€ / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket

Récupel-bijdrage uitgesloten
This item is not returnable. View return policy

Description

Description

Temperature probes are used to measure values in various educational, laboratory, and industrial applications. RTD Pt 100 Temperature Probe comes in various shaft lengths and diameters for use with Temp 16 RTD Thermometer.

Features:

  • Parameter: Temperature
  • Used with: Temp 16 RTD Thermometer
Specifications

Specifications

RTD Pt 100 temperature probe. Shaft length: 500 mm; round-tip sensor: 6 mm diameter; -50 to 400 °C; cable length: 1.5 m
-50°C to +400°C
6 mm
1.5 m
Temp 16 RTD Thermometer
Temperature Probe
500 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.