missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(RS)-AMPA hydrobromide, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Description
More water soluble hydrobromide salt of (RS)-AMPA (Cat. No. 0169).
Specifications
Specifications
Quantity | 1 mg |
Formula Weight | 267.08 |
Percent Purity | >99% |
Chemical Name or Material | (RS)-AMPA hydrobromide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'