missing translation for 'onlineSavingsMsg'
Learn More

RS 23597-190 hydrochloride, Tocris Bioscience™

Product Code. 30001117
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

5-HT4 antagonist

A high affinity, selective competitive antagonist at 5-HT4 receptors.

Chemical Identifiers

CAS 149719-06-2
Molecular Formula C16H24Cl2N2O3
Molecular Weight (g/mol) 363.279
InChI Key QLZLBYYNMGQIAR-UHFFFAOYSA-N
PubChem CID 182592
IUPAC Name 3-piperidin-1-ylpropyl 4-amino-5-chloro-2-methoxybenzoate;hydrochloride
SMILES COC1=CC(=C(C=C1C(=O)OCCCN2CCCCC2)Cl)N.Cl

Specifications

Quantity 10 mg
Synonym 3-piperidin-1-yl propyl 4-amino-5-chloro-2-methoxybenzoate hydrochloride, rs 23597-190 hydrochloride, 3-piperidine-1-yl propyl 4-amino-5-chloro-2-methoxybenzoate hydrochloride, 3-piperidin-1-yl propyl4-amino-5-chloro-2-methoxybenzoate hydrochloride, 3-piperidine-1-yl propyl-4-amino-5-chloro-2-methoxybenzoate hydrochloride, 3-piperidin-1-ylpropyl 4-amino-5-chloro-2-methoxybenzoate hydrochloride, 3-piperidin-1-ylpropyl 4-azanyl-5-chloranyl-2-methoxy-benzoate hydrochloride, 4-amino-5-chloro-2-methoxy-benzoic acid 3-piperidinopropyl ester hydrochloride, 4-amino-5-chloro-2-methoxybenzoic acid 3-1-piperidinyl propyl ester hydrochloride, benzoic acid, 4-amino-5-chloro-2-methoxy-, 3-piperidine-1-yl propyl ester, hydrochloride
Formula Weight 363.28
Percent Purity >98%
Chemical Name or Material RS 23597-190 hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'