missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL26 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517093
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L24P family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
| RPL26 | |
| Polyclonal | |
| Unconjugated | |
| RPL26 | |
| 60S ribosomal protein L26; DBA11; hm:zehn2026; hypothetical protein LOC406387; L26; Large ribosomal subunit protein uL24; ribosomal protein L26; RPL26; SIG-20; silica-induced gene 20 protein; similar to ribosomal protein L26; wu:fa92f05; zgc:66190 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19941, 287417, 6154 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 39 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P12749, P61254, P61255 | |
| RPL26 | |
| Synthetic peptide corresponding to the sequence of human ribosomal protein L26. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction