Learn More
Heidolph instruments Rotavac Vario Tec RPM-Regulated Pump
Use this rpm-regulated rotary evaporator vacuum pump to reach an maintain a precise vacuum level. Heidolph™ Rotavac Vario Tec RPM-Regulated Pump adjusts its speed and runs intermittently at very low speed to hold a constant vacuum.
Brand:  Heidolph instruments 591-00171-00
Product Code. 11701659
  • 3690.00€ / Each
Estimated Shipment: 03-04-2025
to see stock.
Explore more special offers
Add to basket
Récupel-bijdrage uitgesloten
Description
Description
  • Two-stage diaphragm pump is made from chemical-resistant material.
  • Recommended for solvents with low or medium boiling points.
  • Can be combined with a condenser.
  • Speed adjustment stops completely if vacuum is reached and runs intermittently at very low speed to hold vacuum constant.
  • Increased diaphragm performance life due to periods of non-operation.
  • Low energy consumption due to intermittent operation.
  • Significant noise and vibration reduction due to periods of non-operation.
  • Operates without vacuum valve.
  • Precise vacuum level at all times.
Specifications
Specifications
156 x 236 x 196 mm
160 W
IP65
Rotary evaporator vacuum pump
4.3 kg
12 Mbar
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products