missing translation for 'onlineSavingsMsg'
Learn More

Ro 90-7501, Tocris Bioscience™

Product Code. 18084942
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inhibitor of Aβ42 fibril formation

Inhibitor of amyloid β42 (Aβ42) fibril assembly; reduces Aβ42-induced toxicity (EC50 = 2 μM).

Chemical Identifiers

CAS 293762-45-5
Molecular Formula C20H16N6
Molecular Weight (g/mol) 340.39
InChI Key PAGZCEHLFCJSPV-UHFFFAOYSA-N
Synonym gnf-pf-5510, 2'-4-amino-phenyl-3h,3'h-2,5' bibenzoimidazolyl-5-ylamine, 2-2-4-aminophenyl-3h-1,3-benzodiazol-5-yl-3h-1,3-benzodiazol-5-amine, cbchromo1_000149, cambridge id 5180073, ro hplc, 2'-4-aminophenyl-2,5'-bi-1h-benzimidazol-5-amine, 2'-4-aminophenyl-1h,1'h-2,5'-bibenzimidazol-5-amine, 2,5'-bi-1h-benzimidazol-5-amine, 2'-4-aminophenyl, 2-2-4-aminophenyl benzimidazol-6-yl benzimidazole-6-ylamine
PubChem CID 824226
IUPAC Name 2-[2-(4-aminophenyl)-3H-benzimidazol-5-yl]-3H-benzimidazol-5-amine
SMILES C1=CC(=CC=C1C2=NC3=C(N2)C=C(C=C3)C4=NC5=C(N4)C=C(C=C5)N)N

Specifications

Quantity 10 mg
Formula Weight 340.39
Percent Purity >99%
Chemical Name or Material Ro 90-7501
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'