missing translation for 'onlineSavingsMsg'
Learn More

Ro 61-8048, Tocris Bioscience™

Product Code. 18025822
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent kynurenine 3-monooxygenase (KMO) inhibitor

Ro 61-8048 is a potent and competitive kynurenine 3-monooxygenase (kynurenine 3-hydroxylase; KMO) inhibitor (Ki = 4.8 nM, IC50 = 37 nM). Increases kynurenic acid levels to concentrations that antagonize the glycine site of NMDA receptors. Brain penetrant and exhibits antidystonic, anticonvulsant and neuroprotective activities. Ro 61-8048 decreases nicotine self-administration in vivo. Ro 61-8048 prevents post-operative brain edema and consequent neuronal apoptosis in a rat model of surgically induced brain injury.

Chemical Identifiers

CAS 199666-03-0
Molecular Formula C17H15N3O6S2
Molecular Weight (g/mol) 421.442
InChI Key NDPBMCKQJOZAQX-UHFFFAOYSA-N
Synonym 3,4-dimethoxy-n-4-3-nitrophenyl thiazol-2-yl benzenesulfonamide, 3,4-dimethoxy-n-4-3-nitrophenyl-1,3-thiazol-2-yl benzenesulfonamide, 3,4-dimethoxy-n-4-3-nitro-phenyl-thiazol-2-yl-benzenesulfonamide, 7zr, ro hplc, 3,4-dimethoxy-n-4-3-nitrophenyl-1,3-thiazol-2-y, 3,4-dimethoxy-n-4-3-nitrophenyl-2-thiazolyl-benzenesulfonamide, 3,4-dimethoxy-n-4-3-nitrophenyl-2-thiazolyl benzenesulfonamide, 3,4-dimethoxy-n-4-3-nitrophenyl-1,3-thiazol-2-yl benzene-1-sulfonamide
PubChem CID 5282337
ChEBI CHEBI:34953
IUPAC Name 3,4-dimethoxy-N-[4-(3-nitrophenyl)-1,3-thiazol-2-yl]benzenesulfonamide
SMILES COC1=C(C=C(C=C1)S(=O)(=O)NC2=NC(=CS2)C3=CC(=CC=C3)[N+](=O)[O-])OC

Specifications

Quantity 10 mg
Formula Weight 421.45
Percent Purity >99%
Chemical Name or Material Ro 61-8048
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'