missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emma Safety Footwear Ringo Safety Shoes
A versatile shoe with wipe clean uppers and an 'easy clean' sole. Popular with people working in Food and Catering. For peace of mind, an internal toe cap provides resilient protection against falling objects and unforeseen hazards.
116.00€
Specifications
Certifications/Compliance | EN ISO 20345:2011 |
---|---|
Closure Type | Laces |
Color | Black |
Design | Ringo |
Liner Material | Hydro-Tec™ Sanitized Silver |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15938931
|
Emma Safety Footwear
MM73054048 |
48 |
116.00€
1 pair |
Estimated Shipment: 28-03-2025
Log in to see stock. |
|||||
15948931
|
Emma Safety Footwear
MM73054049 |
49 |
116.00€
1 pair |
Estimated Shipment: 28-03-2025
Log in to see stock. |
|||||
Description
- S2-Safety shoe for wet working conditions
- Full grain leather construction
- GripForce™ Easy Twist Outsole
- Hydro-Tec™ Sanitized Silver
Specifications
EN ISO 20345:2011 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
Steel |
Laces | |
Ringo | |
Leather |