Learn More
Hirschmann™ ringcaps™ for Reflotron™ Disposable Capillary Pipettes
Capillary pipettes with capacity line, for the Roche Reflotron™ system.
Brand:  Hirschmann™ 9550532
Product Code. 10771591
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • With ringmark
  • Conformity certified
  • Manufactured from durable DURAN™ glass
  • Lithium-heparinized
  • 32 μL
  • Capillary for Reflotron™ system
  • Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifications
Specifications
Disposable capillary pipettes with ringmark at 32 μL, Lithium heparinized
ringcaps™
Reflotron
Borosilicate Glass
Single-use