Learn More
Brady™ Rigid DuraSleeve™ Vinyl Wire Marking Inserts for M710
Rigid DuraSleeve™ Wire Marking Inserts for use with Brady Printer M710.
Brand:  Brady™ 312002
 View more versions of this product
Product Code. 17858024
  • 53.75€ / Pack of 500
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Brady™ DuraSleeve™ printable inserts create a durable legend
  • Rigid construction of the DuraSleeve™ wire marking system provides inserts that can easily be inserted into a clean carrier
  • Legend can be changed before or after termination
Specifications
Specifications
M7-23x4.4-7696-WT
White
Tape
Wire and Cable Markers
Data and Telecommunications Labeling, Wire and Cable Labeling
1 Label
-40°F to +158°F
0.0098 in.
4.4 x 23 mm
0.91 in.
Non-adhesive
Gloss
Vinyl
Non-sterile
M710, BMP71
500 Labels
-40°C to +70°C
0.249 mm
0.17 in.
23 mm