Learn More
Brady™ Rigid DuraSleeve™ Labels for M710, M610, M611
Rigid DuraSleeve™ Inserts for use with Brady™ Printers M611, M610 and M710.
Brand: Brady™ 312059
This item is not returnable.
View return policy
Description
- Brady™ DuraSleeve™ printable inserts create a durable legend
- Rigid construction of the DuraSleeve™ wire marking system provides inserts that can easily be inserted into a clean carrier
- Legend can be changed before or after termination
Specifications
M6-16x4.4-7696-WT | |
White | |
Tape | |
Wire and Cable Markers | |
Wire and Cable Labeling | |
1 Label | |
-40°F to +158°F | |
0.0098 in. | |
4.4 x 16 mm | |
0.63 in. |
Non-adhesive | |
Gloss | |
Vinyl | |
Non-sterile | |
M610, M710, M611, BMP61, BMP71 | |
250 Labels | |
-40°C to +70°C | |
0.249 mm | |
0.17 in. | |
16 mm |