Learn More
Epredia™ Richard-Allan Scientific™ Iron Stain, Nuclear Red Thiazin stain solution
Product Code. 12583637 Shop All Epredia Products
Click to view available options
Type:
Hydrochloric Acid Solution (20%)
Iron Stain Kit
Nuclear Fast Red
Potassium Ferrocyanide
Unit Size:
125mL
250mL
Each
Product Code. 12583637
Brand: Epredia™ 88015
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Detect the presence of ferric iron (hemosiderin) in histological or hematological preparations with Epredia™ Richard-Allan Scientific™ Iron Stain.
  • Useful in study of iron deficiency anemia, hemochromatosis and hemosiderosis
  • Iron stains blue, nuclei stains pink-red, and cytoplasm stains pale pink
TRUSTED_SUSTAINABILITY
Specifications
Type Nuclear Fast Red