Learn More
Epredia™ HistoGel™ Specimen Processing Gel
Shop All Epredia Products
Click to view available options
Quantity:
1 kit
12 Pk.
144 Pk.
Unit Size:
Each
Pack of 12
Pack of 144
Description
- Penetrates specimen without allowing specimen to escape from gel
- Outperforms other agarose gel mixtures or thromboplastin at a fraction of the cost
- Eliminates unwanted discoloration around specimens on slides
- Remains inside theparaffin block during cutting and virtually unnoticeable during sectioning
Tekniske data
Tekniske data
| Packaging Type | Vial |
| Product Type | Specimen Processing Gel |
| Quantity | 144 Pk. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion