Learn More
Labconco™ ReVo TUV Stand Caster Kit
The ReVo TUV Stand Caster Kit provides molibity for the ReVo fixed-leg stands.
Brand:  Labconco™ R3730500
Product Code. 17800764
  • 760.00€ / Each
Explore more special offers
Description
Description
  • Provides mobility for ReVo fixed-leg stands.
  • Includes four wheels.
  • Installation required.
  • Select the caster kit that matches your base stand type.
  • Casters make base stands portable for relocation, repositioning and easy cleaning.
Specifications
Specifications
H14 HEPA
4 Wheels
130 x 130 x 60 mm
1 kg