Learn More
Labconco™ ReVo MSC IV Bar
Bar supports intravenous solution bottles and bags. Kit includes IV bar and mounting hardware.
Brand:  Labconco™ R3858600
 View more versions of this product
Product Code. 17825602
  • 1250.00€ / 1 unit
Explore more special offers
Description
Description
  • IV Bar and mounting hardware compatible with various biosafety cabinets, including Purifier Axiom Biosafety Cabinets, Purifier Cell Logic and Cell Logic+ Biosafety Cabinets, Purifier Logic and Logic+ Biosafety Cabinets, Purifier Logic and PuriCare Procedure Stations and ReVo Microbiological Safety Cabinets.
Specifications
Specifications
IV Bag Holder Kit
900 x 20 x 20 mm
2.0 kg