Learn More
Labconco™ ReVo MSC IV Bar
Bar supports intravenous solution bottles and bags. Kit includes IV bar and mounting hardware.
Brand: Labconco™ R3858603
This item is not returnable.
View return policy
Description
- IV Bar and mounting hardware compatible with various biosafety cabinets, including Purifier Axiom Biosafety Cabinets, Purifier Cell Logic and Cell Logic+ Biosafety Cabinets, Purifier Logic and Logic+ Biosafety Cabinets, Purifier Logic and PuriCare Procedure Stations and ReVo Microbiological Safety Cabinets.
Specifications
IV Bag Holder Kit | |
1800 x 20 x 20 mm | |
3.0 kg |