Learn More
Labconco™ ReVo Canopy Connection Kit
ReVo Canopy Connection Kit is ideal for ducting ReVo Class II, Type A2 Microbiological Safety Cabinets.
1305.00€ - 1410.00€
Specifications
Product Type | Canopy Connection Kit |
---|---|
Dimensions (L x W x H) Exterior | 465 x 680 x 265 mm |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17885592
|
Labconco™
R3889210 |
900/1200 mm Cabinet |
1305.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17875592
|
Labconco™
R3889212 |
1500/1800 mm Cabinet |
1410.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Canopy Connection Kit with side exhaust for ducting ReVo Class II, Type A2 Microbiological Safety Cabinets.
Specifications
Canopy Connection Kit | |
465 x 680 x 265 mm |