Learn More
Labconco™ ReVo Canopy Connection Kit
ReVo Canopy Connection Kit is ideal for ducting ReVo Class II, Type A2 Microbiological Safety Cabinets.
Brand: Labconco™ R3889202
This item is not returnable.
View return policy
Description
- Canopy Connection Kit with top exhaust for ducting ReVo Class II, Type A2 Microbiological Safety Cabinets.
Specifications
Canopy Connection Kit | |
465 x 680 x 265 mm | |
1500/1800 mm Cabinet | |
5.0 kg |