Learn More
Labconco™ ReVo Canopy Connection Kit
ReVo Canopy Connection Kit is ideal for ducting ReVo Class II, Type A2 Microbiological Safety Cabinets.
Brand:  Labconco™ R3889200
 View more versions of this product
Product Code. 17805602
  • 1305.00€ / 1 set
Explore more special offers
Description
Description
  • Canopy Connection Kit with top exhaust for ducting ReVo Class II, Type A2 Microbiological Safety Cabinets.
Specifications
Specifications
Canopy Connection Kit
465 x 680 x 265 mm
900/1200 mm MSC
5.0 kg