Learn More
Brady™ Retro Reflective All Weather Label Tape for M6/M7 Printers
Product Code. 17808744
Click to view available options
Color:
Orange
Silver
Yellow
Dimensions (LxW):
30 ft. x 1 in.
30 ft. x 2 in.
Unit Size:
1 roll
Each
Product Code. 17808744
Brand: Brady™ 174128
Only null left
Add to basket
Add to basket
Highly reflective material shines brightly when struck by light which is ideal for applications where high visibility is critical.
  • Ideal for labeling job site tools, hard hats, utility enclosures, poles and pedestals, rooftop equipment, racks and shelves
  • Retroreflective sheeting and pressure sensitive acrylic adhesive meet Federal Spec L-S-300 for Reflectivity
  • Average outdoor durability is 4 to 6 years
  • Service temperature of -40°C to 70°C (-40°F to 158°F)
  • Made of B-584 plastic material
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance ASTM D 2979, Fed. Spec. L-S-300 (Class 1)
Adhesive Type Permanent Acrylic
Coating None
Color Orange
Finish Gloss
Format Continuous
Material Plastic
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Component and Equipment Labeling, Facility Identification, General Identification, Safety Identification
For Use With (Equipment) M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
Latex Free No
Quantity 30 ft.
Shape Rectangle
Temperature Range (English) -40°F to +158°F
Temperature Range (Metric) -40°C to +70°C
Thickness (English) 0.0055 in.
Thickness (Metric) 0.14 mm
Dimensions (LxW) 30 ft. x 2 in.
Length (English) 30 ft.
Length (Metric) 9.14 m
Width (English) 2 in.
Width (Metric) 50.8 mm
Alternate Part Number M6C-2000-584-OR