missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Replacement Pump Ball for Buerkle™ Solvent Pump
Buerkle™ Replacement Pump Ball for use with Buerkle™ Hand Operated Solvent Pump.
Brand: Buerkle™ 5603-0015
102.77 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
- Spare part
- For use with Buerkle™ Hand Operated Solvent Pump
Specifications
1 Ea. | |
Replacement Pump Balls | |
Solvent Pump |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction