Learn More
Grant Instruments™ Polycarbonate Transparent Lid, Gabled
Product Code. 11476417
Click to view available options
For Use With (Equipment):
26L Tanks
2L Tanks
5L Tanks. Fits JBA5/JBN5 bath
SAP12, JBN12, JBA12, SBB Aqua 12+
Unit Size:
Each
Product Code. 11476417
Brand: Grant Instruments™ AQL2
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Directs condensation away from immersed vessels, avoids contamination, reduces evaporation and saves energy.
  • Polycarbonate construction
  • Clear, gabled lid allows condensation buildup to drip away from immersed vessels.
  • For use with Sub Aqua Pro Series Unstirred Water Baths
TRUSTED_SUSTAINABILITY
Specifications
Description Gabled
For Use With (Equipment) 2L Tanks
Material Polycarbonate