Learn More
Thermo Scientific™ Remel™ Tryptic Soy Broth
The Thermo Scientific™ Remel Tryptic Soy Broth (TSB) is a general purpose enrichment broth for cultivation and isolation of microorganisms.
203.97€
Product Code | Brand | Format | Packaging Type | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Packaging Type | Quantity | Price | Quantity & Availability | |||
17719448
|
Thermo Scientific™
R112745 |
BB Septum - 100mL | 10/Pk. | 10 x 100 mL |
203.97€
Pack of 10 |
Estimated Shipment: 20-11-2024
Log in to see stock. |
|||
Description
Thermo Scientific™ Remel Tryptic Soy Broth (TSB) is a general purpose enrichment broth for cultivation and isolation of microorganisms.
- Part numbers beginning with R11 are for sterility testing according to USP (United States Pharmacopeia) and for use as general purpose enrichment broth
Specifications
Tryptic Soy Broth | |
Microbiology Media |
REF numbers beginning with R11 are not intended for IVD use.