missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Shrink-Seal

Thermo Scientific™ Remel Shrink-Seal is used to seal 100mL petri dishes.

Brand:  Thermo Scientific™ R522600

Product Code. 10301793

  • 139.00€ / Pack of 144
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Remel Shrink-Seal is used to seal 100mL petri dishes.

  • Shrinks as it dries
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sealing 100 mL Petri Dishes
Shrinks as it dries; 144/Pk.
Shrink-Seal
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions