Learn More
Olympus™ Reflected Light Illuminator, Brightfield
Compact and efficient, the Olympus™ Reflected Light Illuminator, Brightfield is conveniently sized but offers 100W halogen illumination.
Brand:  Olympus™ K10017494
Product Code. 12550206
  • 2150.00€ / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For brightfield illumination.
  • Compact with reduced depth.
  • Halogen fiber illumination.
Specifications
Specifications
Halogen
Brightfield
100 w