Learn More
Labcon Refillable Racks for Labcon Tips
GREENER_CHOICE
Product Code. 16681963
Click to view available options
Holds:
Tips (10 to 1250μL)
Tips (Labcon 10μL)
Unit Size:
Pack of 10
Pack of 12
Product Code. 16681963
Brand: Labcon 12999600369
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
For use with Eclipse™ pipette tip refills
  • Pipette tip rack
  • Refillable
  • With hinged clear lid
  • Made in USA
Specifications
Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Product Type Empty Refill Rack with Lid
Autoclavable Autoclavable
Color Purple
Holds Tips (Labcon 10μL)
Material Polypropylene
Positions Holds one refill platform of 96 tips
Sterility Non-sterile
For Use With (Application) Liquid Handling
Packaging 12 racks / Pk./10 Pk. / case
Quantity 12 racks / Pk.
Shipping Condition RT
Size 10 μL tips