Learn More
WTW™ Reference Electrode
Reference electrodes are necessary if you are working with half cells during ISE measurement. They provide the reference potential and can be combined with any ISE half cells. The bridge electrolyte can be exchanged easily, depending on the application.
Brand:  WTW™ 106571
Product Code. 10051292