missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Streptolysin Protein
Recombinant Protein
1295.00€ - 2050.00€
Specifications
Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4. |
---|---|
Molecular Weight (g/mol) | MolecularWeight-theroretical: 60.1 kDa |
Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
Endotoxin Concentration | Less than 0.1 EU/μg of Streptolysin as determined by LAL method. |
Gene Alias | Slo, Thiol activated cytolysin |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18634946
|
Novus Biologicals™
|
1 mg |
2050.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18688668
|
Novus Biologicals™
NBP2-61445-500ug |
500 μg |
1295.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Data Not Available.Specifications
Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4. | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) | |
Slo, Thiol activated cytolysin | |
>97 % pure by SDS-PAGE and HPLC |
MolecularWeight-theroretical: 60.1 kDa | |
Less than 0.1 EU/μg of Streptolysin as determined by LAL method. | |
Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) | |
Streptolysin |