Learn More
R&D Systems™ Recombinant Rat Lipocalin-2/NGAL Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand:  R&D Systems™ 3508-LC-050
Product Code. 18137230
  • 394.00€ / 50µg
Explore more special offers
Description
Description
The Recombinant Rat Lipocalin-2/NGAL Protein is derived from NS0. The Recombinant Rat Lipocalin-2/NGAL Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in MES and NaCl.
21 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
170496
Lipocalin-2/NGAL
Mouse myeloma cell line,NS0-derived rat Lipocalin-2/NGAL protein Gln21-Asn198,with a C-terminal 6-His tag
24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin2, Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGAL, NGALlipocalin-2, Oncogene 24p3, p25, Siderocalin, Uterocalin
Recombinant
Product Suggestions