missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other T. pallidum p47 Protein
A cDNA sequence encoding the T. pallidum p47 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13675-500ug
This item is not returnable.
View return policy
Specifications
T. pallidum p47 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
8M urea, 10mM Tris-HCl pH 8.0, 1mM EDTA and 1mM DTT. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Treponema Pallidum protein is >95% pure as determined by SDS-PAGE. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction