missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Renin Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 4277-AS-020

Product Code. 18137510

  • 362.00€ / 20µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its ability to cleave the fluorogenic peptide substrate 5-FAM/QXL™ 520. Peti-Peterdi, J. et al. (2009) Physiology 24:88. The specific activity is >20 pmol/min/μg, as measured under the described conditions.
Specifications

Specifications

P06281
Supplied as a 0.2 μm filtered solution in MES and NaCl.
M.W. (Observed): 45-55 kDa, reducing conditions; M.W. (theoretical): 43 kDa
20 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Ren1
Recombinant
Renin
Enzyme Activity
5972
Renin
Mouse myeloma cell line, NS0-derived mouse Renin protein Leu22-Arg402, with a C-terminal 10-His tag
<1.0 EU / 1 μg of the protein by the LAL method.
Unconjugated
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.