Learn More
R&D Systems™ Recombinant Mouse Plexin A2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 5486-PA-050
Product Code. 18188570
  • 394.00€ / 50µg
Explore more special offers
Description
Description
The Recombinant Mouse Plexin A2 Protein is derived from CHO. The Recombinant Mouse Plexin A2 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
133 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
18845
Plexin A2
Chinese Hamster Ovary cell line,CHO-derived mouse Plexin A2 protein Met25-Pro1227,with a C-terminal 6-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.