Learn More
R&D Systems™ Recombinant Mouse Lymphotoxin alpha2/beta1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 1008-LY-010/CF
307.55 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18107630
  • 390.00€ / 10µg
Explore more special offers
Description
Description
The Recombinant Mouse Lymphotoxin alpha2/beta1 Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse Lymphotoxin alpha2/beta1 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
18.5 kDa (LT alpha), 18.4 kDa (LTbeta)
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
16992
Lymphotoxin α2/β1
Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse Lymphotoxin protein Leu34-Leu202 (LT alpha ) & Gly136-Gly306 (LT beta )
Noncovalently-linked heterotrimer
Recombinant
Product Suggestions