Learn More
R&D Systems™ Recombinant Mouse Integrin alpha V beta 6 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 7480-AV-050
Product Code. 18148110
  • 543.00€ / 50µg
Explore more special offers
Description
Description
The Recombinant Mouse Integrin alpha V beta 6 Protein is derived from CHO. The Recombinant Mouse Integrin alpha V beta 6 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
Integrin alpha V beta 6
Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha V beta 6 protein, (N-terminus) Mouse Integrin alpha V (Phe31-Val988) Accession &Num; P43406 HP GGGSGGGSGGGS Acidic Tail HHHHHH Mouse Integrin beta 6 (Gly22-Asn706) Accession &Num; Q9Z0T9 GGGSGGGSGGGS Basic Tail (C-terminus)
Non-covalently-linked heterodimer
Recombinant
115 kDa ( alphaV subunit) & 82.6 kDa (beta6 subunit)
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.