missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Integrin alpha M beta 2 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 7959-AM-050

433.68 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18186240

  • 559.00€ / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Mouse Integrin alpha M beta 2 Protein is derived from CHO. The Recombinant Mouse Integrin alpha M beta 2 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
Integrin alpha M beta 2
Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha M beta 2 protein, (N-terminus) Mouse Integrin alpha M (Phe17-Asn1105) Accession &Num; NP_032427 His-Pro GGGSGGGS Acidic Tail 6-His tag Mouse Integrin beta 2 (Gln24-Asn702) Accession &Num; P11835 His-Pro GGGSGGGS Basic Tail (C-terminus)
Noncovalently-linked heterodimer
Recombinant
130 kDa (Integrin alphaM) & 83 kDa (Integrin beta2)
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE with silver staining.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions