missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Hemoglobin subunit beta-2 Protein

A DNA sequence encoding the Mus musculus (Mouse) Hemoglobin subunit beta-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7346-ec-50ug

 View more versions of this product

Product Code. 16018360

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_058652.1
Hemoglobin subunit beta-2 Protein
Research Use Only
Hbb-b2
Recombinant Protein
E. coli
VHLTDAEKSA VSCLWAKVNP DEVGGEALGR LLVVYPWTQR YFDSFGDLSS ASAIMGNPKV KAHGKKVITA FNEGLKNLDN LKGTFASLSE LHCDKLHVDP ENFRLLGNAI VIVLGHHLGK DFTPAAQAAF QKVVAGVATA LAHKYH
Greater than 90% as determined by SDS-PAGE.
15130
50 μg
Not determined.
Full Length
Mouse
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.