missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse HAI-2B Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 1107-PI-010

307.55 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18187400

  • 390.00€ / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Mouse HAI-2B Protein is derived from NS0. The Recombinant Mouse HAI-2B Protein has been validated for the following applications: Inhibition Activity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
14 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
20733
HAI-2B
Mouse myeloma cell line,NS0-derived mouse HAI-2B protein Arg30-Lys140,with a C-terminal 10-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions