missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Glycoprotein V/CD42d Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 6990-GP-050
This item is not returnable.
View return policy
Description
The Recombinant Mouse Glycoprotein V/CD42d Protein is derived from NS0. The Recombinant Mouse Glycoprotein V/CD42d Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
57.5 kDa (monomer) | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
14729 | |
Glycoprotein V/CD42d | |
Mouse myeloma cell line,NS0-derived mouse Glycoprotein V/CD42d protein Met1-Gly522 with a C-terminal 6-His tag | |
Dimer | |
Recombinant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction