missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse FCRL5/FcRH3 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 6757-FC-050

319.65 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18117210

  • 407.00€ / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Mouse FCRL5/FcRH3 Protein is derived from NS0. The Recombinant Mouse FCRL5/FcRH3 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
53.5 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
329693
FCRL5/FcRH5
Mouse myeloma cell line,NS0-derived mouse FCRL5/FcRH5 protein Met1-Ala496 with a C-terminal 6-His tag
Unconjugated
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions