Learn More
R&D Systems™ Recombinant Mouse Carbonic Anhydrase XIV Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 2504-CA-010
Product Code. 18107210
  • 292.65€ / 10µg
Explore more special offers
Description
Description
The Recombinant Mouse Carbonic Anhydrase XIV Protein is derived from NS0. The Recombinant Mouse Carbonic Anhydrase XIV Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Supplied as a 0.2μm filtered solution in Tris and NaCl.
32 kDa
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Recombinant
23831
Carbonic Anhydrase XIV/CA14
Mouse myeloma cell line,NS0-derived mouse Carbonic Anhydrase XIV/CA14 protein Ala16-Met290,with a C-terminal 10-His tag Accession &Num; Q9WVT6
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.