missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Monkey GM CSF Protein

Product Code. 15994508
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15994508

missing translation for 'mfr': enQuireBio™ QP1064710ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the GM CSF was constructed and used to recombinantly synthesize the protein.

Specifications

Name Monkey GM CSF Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 0.1ng/ml, corresponding to a specific activity of > 10,000,000 IU/mg.
Product Type Recombinant Protein
Cross Reactivity Donkey
Species E. coli
Protein Tag Untagged
Sequence APARSPSPGT QPWEHVNAIQ EARRLLNLSR DTAAEMNKTV EVVSEMFDLQ EPSCLQTRLE LYKQGLQGSL TKLKGPLTMM ASHYKQHCPP TPETSCATQI ITFQSFKENL KDFLLVIPFD CWEPVQE
Buffer GM-CSF was lyophilized from a concentrated (1 mg/ml) solution containing 1x PBS pH 7.4.
Purity or Quality Grade Greater than 98.0% as determined by SDS-PAGE and RP-HPLC.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.