Learn More
R&D Systems™ Recombinant Human Uteroglobin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 4218-UT-050
This item is not returnable.
View return policy
Description
The Recombinant Human Uteroglobin Protein is derived from E. coli. The Recombinant Human Uteroglobin Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
8 kDa (monomer) | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
7356 | |
Uteroglobin/SCGB1A1 | |
E. coli-derived human Uteroglobin/SCGB1A1 protein Glu22-Asn91,with an N-terminal Met | |
Disulfide-linked homodimer | |
Recombinant |
Product Content Correction