missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TSNAX Protein
A cDNA sequence encoding the TSNAX was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13829-1mg
This item is not returnable.
View return policy
Specifications
TSNAX Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSNKEGS GGFRKRKHDN FPHNQRREGK DVNSSSPVML AFKSFQQELD ARHDKYERLV KLSRDITVES KRTIFLLHRI TSAPDMEDIL TESEIKLDGV RQKIFQVAQE LSGEDMHQFH RAITTGLQEY VEAVSFQHFI KTRSLISMDE INKQLIFTTE DNGKENKTPS SDAQDKQFGT WRLRVTPVDY LLGVADLTGE LMRMCINSVG NGDIDTPFEV SQFLRQVYDG FSFIGNTGPY EVSKKLYTLK QSLAKVENAC YALKVRGSEI PKHMLADVFS VKTEMIDQEE GIS | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TSNAX protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.2M NaCl, 50% glycerol and 1mM DTT. |