missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human TRANCE/RANK L/TNFSF11 (E. coli-expressed)

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

480.00€

Specifications

Gene ID (Entrez) 8600
Name TRANCE/TNFSF11/RANK L
Quantity 10 μg
Source E. coli-derived human TRANCE/TNFSF11/RANK L protein Arg85-Asp245,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
View More Specs

Products 2
Product Code Brand Formulation Price Quantity & Availability  
Product Code Brand Formulation Price Quantity & Availability  
18146710
View Documents
R&D Systems™
6449-TEC-010
Lyophilized from a 0.2μm filtered solution in MES and NaCl with BSA as a carrier protein.
480.00€
10µg
Please to purchase this item. Need a web account? Register with us today!
18188030
View Documents
R&D Systems™
6449-TEC-010/CF
Lyophilized from a 0.2μm filtered solution in MES and NaCl.
480.00€
10µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Recombinant Human TRANCE/RANK L/TNFSF11 (E. coli-expressed) is derived from E. coli. The Recombinant Human TRANCE/RANK L/TNFSF11 (E. coli-expressed) has been validated for the following applications: Bioactivity.
Specifications

Specifications

8600
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
TRANCE/TNFSF11/RANK L
E. coli-derived human TRANCE/TNFSF11/RANK L protein Arg85-Asp245,with an N-terminal Met
Unconjugated
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.