missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFRSF6B Protein
A cDNA sequence encoding the TNFRSF6B was constructed and used to recombinantly synthesize the protein.
143.00€ - 4090.00€
Specifications
Gene ID (Entrez) | 8771 |
---|---|
Name | TNFRSF6B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | TNFRSF6B |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15924649
|
enQuireBio™
QP13770-5ug |
5 μg |
143.00€
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15914649
|
enQuireBio™
QP13770-20ug |
20 μg |
222.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15904649
|
enQuireBio™
QP13770-1mg |
1 mg |
4090.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
8771 | |
Research Use Only | |
TNFRSF6B | |
Human | |
His | |
TNFRSF6B protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
TNFRSF6B Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSVAETPTY PWRDAETGER LVCAQCPPGT FVQRPCRRDS PTTCGPCPPR HYTQFWNYLE RCRYCNVLCG EREEEARACH ATHNRACRCR TGFFAHAGFC LEHASCPPGA GVIAPGTPSQ NTQCQPCPPG TFSASSSSSE QCQPHRNCTA LGLALNVPGS SSHDTLCTSC TGFPLSTRVP GAEECERAVI DFVAFQDISI KRLQRLLQAL EAPEGWGPTP RAGRAALQLK LRRRLTELLG AQDGALLVRL LQALRVARMP GLERSVRERF LPVH | |
Greater than 85% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title