missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Thioredoxin-1 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Brand:  R&D Systems™ 1970-TX-500

Product Code. 18604749

  • 507.00€ / 500µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its ability to catalyze the reduction of insulin. The reaction leads to precipitation, which can be measured by absorbance at 650 nm. The specific activity is >10 A650/min/mg, as measured under the described conditions.
Specifications

Specifications

P10599
Lyophilized from a 0.2 μm filtered solution in PBS, EDTA and DTT.
M.W. (Observed): 12 kDa, reducing conditions; M.W. (theoretical): 12 kDa
500 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
TXN
Recombinant
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Enzyme Activity
7295
Thioredoxin-1
E. coli-derived human Thioredoxin-1 protein Val2-Val105
<0.10 EU / 1 μg of the protein by the LAL method.
Unconjugated
Reconstitute at 1000 μg/mL in sterile PBS.
Thioredoxin-1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.