Learn More
R&D Systems™ Recombinant Human SULT2A1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ 5828-ST-020
This item is not returnable.
View return policy
Description
The Recombinant Human SULT2A1 Protein is derived from E. coli. The Recombinant Human SULT2A1 Protein has been validated for the following applications: Enzyme Activity.Specifications
Supplied as a 0.2μm filtered solution in Tris, NaCl and EDTA. | |
35 kDa | |
20 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. | |
Recombinant |
6822 | |
Cytosolic Sulfotransferase 2A1/SULT2A1 | |
E. coli-derived human Cytosolic Sulfotransferase 2A1/SULT2A1 protein Ser2-Glu285,with an N-terminal Met and 6-His tag | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |