missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SIX6 Protein

A cDNA sequence encoding the SIX6 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13514-1mg

 View more versions of this product

Product Code. 15913819

  • 4090.00€ / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

SIX6 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMFQLPIL NFSPQQVAGV CETLEESGDV ERLGRFLWSL PVAPAACEAL NKNESVLRAR AIVAFHGGNY RELYHILENH KFTKESHAKL QALWLEAHYQ EAEKLRGRPL GPVDKYRVRK KFPLPRTIWD GEQKTHCFKE RTRNLLREWY LQDPYPNPSK KRELAQATGL TPTQVGNWFK NRRQRDRAAA AKNRLQQQVL SQGSGRALRA EGDGTPEVLG VATSPAASLS SKAATSAISI TSSDSECDI
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SIX6 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 40% glycerol, 5mM DTT and 2mM EDTA.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions