missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Pref-1/DLK1/FA1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
387.00€
Specifications
Gene ID (Entrez) | 8788 |
---|---|
Molecular Weight (g/mol) | 29.7 kDa |
Name | Pref-1/DLK-1/FA1 |
Quantity | 25 μg |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18310258
|
R&D Systems™
1144-PR-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
387.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18128470
|
R&D Systems™
1144-PR-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
387.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human Pref-1/DLK1/FA1 Protein is derived from NS0. The Recombinant Human Pref-1/DLK1/FA1 Protein has been validated for the following applications: Bioactivity.Specifications
8788 | |
Pref-1/DLK-1/FA1 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
29.7 kDa | |
25 μg | |
Unconjugated |