Learn More
R&D Systems™ Recombinant Human Plexin A4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 5856-PA-050
Product Code. 18177750
  • 565.00€ / 50µg
Explore more special offers
Description
Description
The Recombinant Human Plexin A4 Protein is derived from CHO. The Recombinant Human Plexin A4 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
135.9 kDa (monomer)
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
91584
Plexin A4
Chinese Hamster Ovary cell line,CHO-derived human Plexin A4 protein Thr24-Pro1237,with a C-terminal 6-His tag
Noncovalently-linked homodimer
Recombinant
Product Suggestions