Learn More
R&D Systems™ Recombinant Human Plexin A4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 5856-PA-050
This item is not returnable.
View return policy
Description
The Recombinant Human Plexin A4 Protein is derived from CHO. The Recombinant Human Plexin A4 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
135.9 kDa (monomer) | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
91584 | |
Plexin A4 | |
Chinese Hamster Ovary cell line,CHO-derived human Plexin A4 protein Thr24-Pro1237,with a C-terminal 6-His tag | |
Noncovalently-linked homodimer | |
Recombinant |
Product Content Correction